Learn More
Serim Research Corp Serim MONITOR™ for Chlorine 0 - 10 ppm Test Strips
SDP
Supplier:  Serim Research Corp 5155
Encompass_Preferred
Test Measures free chlorine concentrations of 0 - 10 ppm. Useful for multiple applications and industries. Sold as one bottle of 100 tests
Catalog No. NC0924071
Explore available promotions