Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SeraCare™ Anti-HCV Mixed Titer Performance Panel
Specimens are undiluted aliquots from plasma units collected from asymptomatic blood donors from 1998 to 2007
Supplier: SeraCare™ 08100144
Description
- Package of 25 members, 23 members with anti-HCV reactivities ranging from weak to strong positive and two members negative for anti-HCV
- For use by manufacturers and laboratories to evaluate their Anti-HCV test systems using well-characterized specimens and to provide comprehensive data for comparative analysis
- Units were processed by filtration
- No preservatives
- For research use only: Not for use in diagnostic procedures
Specifications
-10°C | |
Anti-HCV Mixed Titer Performance Panel | |
Evaluating Anti-HCV test system using well characterized specimens, to provide comprehensive data for comparative analysis |
Panel | |
Set of 25 specimens from Plasma units with reactivities ranging from weak to strong positive for Anti-HCV | |
25 x 1 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction