Learn More
Thermo Scientific™ Sensititre™ARIS HiQ™ System for AST
Achieve comprehensive, accurate MIC results to guide targeted patient therapy with this benchtop, automated reading and incubation system for AST.
Supplier: Thermo Scientific™ SW4000
Description
Achieve the comprehensive, accurate MIC values needed to guide more targeted patient therapy with the Thermo Scientific™ Sensititre™ ARIS HiQ™ System, a benchtop automated incubation and reading system for antimicrobial susceptibility testing and identification of bacterial pathogens; designed to accommodate up to 100 Sensititre™ microtitre plates within a single instrument.
Confidently detect emerging antibiotic resistance with the gold-standard level accuracy1,2 of broth microdilution coupled with time-saving benefits of automation to optimize patient care, support antimicrobial stewardship, and improve lab efficiency with the Sensititre™ ARIS HiQ™ System.
- Efficient plate handling: central turntable for loading and unloading Sensititre plates via robotic plate transfer and advanced sorting options to optimize read times and efficient plate removal
- Flexible loading: batch load and unload capability for improved testing workflow
- Seamless LIS connectivity: Thermo Scientific™ Sensititre SWIN™ software connection is configured to meet LIS connectivity requirements
- Large capacity, small footprint: to process more tests at one time with limited impact to bench space.
The Sensititre ARIS HiQ system helps microbiology laboratories deliver accurate antimicrobial susceptibility testing of the most commonly prescribed antibiotics, as well as novel, last-resort therapies, to provide meaningful results for clinicians, stewardship teams, and public health departments. By providing critical data to support decisions on prescribing the right antibiotic, at the right dose, at the right time, the Sensititre AST System supports the effort to improve patient outcomes and to ensure the right actions are taken to protect the future of healthcare, veterinary, and agriculture industries against the threat of antibiotic resistance.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
SWIN PC Kit Intl | |
Microbiology Media | |
Minimum Inhibitory Concentration (MIC) |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Thermo Fisher Scientific products are distributed globally so uses, applications, and availability of product in each country depend on local regulatory marketing authorization status.