Learn More
Electron Microscopy Sciences Shorter Pin SEM Stub Slotted 1/2" Head with Shorter Pin (6 mm), Aluminum, 1/8" Pin diameter
Supplier: Electron Microscopy Sciences 7522510
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Cambridge, Phillips, Camscan, B&L, Etec, Zeiss SEM specimen mounts. Tapered end pin, slotted head.
Head: 1/2"
Pin height: 6 mm
Pin diameter: 1/8"