Learn More
Electron Microscopy Sciences Standard Pin SEM Stub 90° Double, 1" Head with 3/8" Pin, Aluminum
Supplier: Electron Microscopy Sciences 75348
Double 90° SEM specimen mount with aluminum slotted head for use with FEI, Tescan, Zeiss, Philips, LEO, Cambridge, AMRAY, Leica, CamScan, ETEC. Made from ultra-pure aluminum.
Head: 1"
Pin height: 3/8"