Learn More
Electron Microscopy Sciences Standard Pin SEM Stub 70° Low Profile, 1/2" Head with 3/8" Pin, Aluminum
Supplier: Electron Microscopy Sciences 75354
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
70° low profile SEM specimen mount for use with FEI, Tescan, Zeiss, Philips, LEO, Cambridge, AMRAY, Leica, CamScan, ETEC. Made from ultra-pure aluminum.
Head: 1/2"
Pin height: 3/8"