Learn More
Electron Microscopy Sciences Standard Pin SEM Stub 100 mm Head, Aluminum, 3.2 mm Pin diameter
Supplier: Electron Microscopy Sciences 7523910
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Pin stub mount for use with AMRAY, Cambridge, Leica, ZEISS/LEO, FEI/PHILIPS, CanScan, and Tescan. Made from ultra-pure aluminum with a standard lathe finish.
Head: 100 mm
Pin diameter: 3.2 mm