Learn More
Electron Microscopy Sciences Standard Pin SEM Stub Mini, 6.6 x 1.3 mm Head, 9.5 mm Pin, Aluminum
Supplier: Electron Microscopy Sciences 7563820
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Mini SEM pin stub mount for Cambridge, Leica, ZEISS/LEO, FEI/Philips, CamScan, Tescan, ETEC and Amray. Made from lathe finished ultra-pure aluminum.
Head: 6.6 x 1.3 mm
Pin height: 3/8"