Description
- The NFPA 1852 Standard on Selection, Care, and Maintenance of Open-Circuit Breathing Apparatus and all major SCBA manufacturers require that compliant SCBAs be flow tested annually on a calibrated breathing machine and by a certified inspector.
- Fisher Scientific service technicians undergo regular training and certification by 3M Scott Fire & Safety, MSA, and Dräger.
- They use the most advanced equipment available, like Honeywell Biosystems PosiChek 3™ test equipment and the OHD Quantifit™ tester to ensure SCBAs are operating as defined by the manufacturer while meeting NFPA standards and OSHA 1910.134 requirements.
- Unlike basic inspections performed prior to each use, a functional SCBA test is conducted against specialized equipment by an approved and certified technician.
- Experienced staff offers these SCBA services
- Flow testing
- Overhauls and upgrades
- Communication systems repair/installation
- General SCBA repairs
- Fit testing using the PortaCount™ respiratory fit tester
Specifications
Safety Equipment and Instrument Services | |
OSHA 29 CFR |