Learn More
SECO™ Rhino Shield™ 510 Foil Bags
$98.93
Catalog Number | Mfr. No. | Length (English) | Width (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Width (English) | Price | Quantity & Availability | ||||
19-045-628
![]() |
SECO™
51068 |
8 in. | 6 in. |
Pack of 100 for $98.93
|
|
||||
19-045-630
![]() |
SECO™
5101215 |
15 in. | 12 in. | N/A | N/A | ||||
Please call customer service for current pricing. \n1-800-766-7000 | |||||||||
Description
The combination of polyethylene for heat sealing and foil for vacuum retention produces a laminated-film material with superior barrier properties against water vapor and oxygen. Flexible, with excellent tensile strength, and protect against atmospheric changes as well as water, grease, acid, chemicals, and corrosion.
Order Info
Available in three sizes. Packaged 100 per pack.
Specifications
Laminated Polyethylene, Foil |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.