Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SECO™ Anti-Static Foil Barrier Bag
The benefits of opaque poly/foil packaging with static dissipative protection
Supplier: SECO™ 5122024CL
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
This item is not returnable.
View return policy
Description
- Used for extremely critical or sensitive devices
- Color: silver/silver
- Size: 20 x 24 in.
- Thickness: 0.0036mil
- Meets the military specifications of MIL-PRF-81705D
Specifications
Foil, Polymer | |
20 in. | |
24 in. |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction