Learn More
Fritsch Milling And Sizing Inc Screw-On Lid Agate, P-5 Pl,125-420 Ml
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Fritsch Milling And Sizing Inc 50.6405.00
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
With the screw-on grinding bowl lid made of agate, you can seal the agate grinding bowl with 420 ml, 250 ml and 125 ml volume of the PULVERISETTE 5 premium line gas-tight for transport between filling in the glove box and the mill.