Learn More
Fritsch Milling And Sizing Inc Screw-On Lid Zirconium Oxide P-5 Pl150-500Ml
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Fritsch Milling And Sizing Inc 50.6495.00
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
With the screw-on grinding bowl lid made of zirconium oxide, you can seal the zirconium oxide grinding bowl with 500 ml, 250 ml and 150 ml volume of the PULVERISETTE 5 premium line gas-tight for transport between fi lling in the glove box and the mill.