Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chembio SCoV-2 Ag Detect™ Rapid Test Kit
![SDP](/content/dam/fishersci/glyphs/sdp.png)
The SCoV-2 Ag Detect Rapid Test Kit can be used for testing symptomatic or asymptomatic patients using a direct nasal sample that gives results in about 20 minutes.
$837.00
Specifications
Content And Storage | 15°C to 30°C |
---|---|
CE Marker | Yes |
Clia Complexity | Waived |
Detectable Analytes | SARS-CoV-2 Antigens |
Sample Type | Direct Nasal Swab |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-999-116
![]() |
Chembio
9020218 |
50 Tests/Kit |
Pack of 50 for $837.00
|
|
|||||
23-999-117
![]() |
Chembio
9120218 |
20 Tests/Kit | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
SCoV-2 Ag Detect™ Rapid Test is a lateral flow immunoassay intended for the qualitative detection of SARS-CoV-2 Nucleoprotein antigen in direct anterior nasal swab specimens from individuals who are suspected of COVID-19 by their healthcare provider within 5 days of symptom onset or from individuals without symptoms or other epidemiological reasons to suspect COVID-19 when tested twice over two or three days with at least 24 hours and no more than 48 hours between tests. Testing is limited to laboratories certified under the Clinical Laboratory Improvement Amendments of 1988 (CLIA), 42 U.S.C. §263a, that meet the requirements to perform moderate, high or waived complexity tests. This test is authorized for use at the Point of Care (POC), i.e., in patient care settings operating under a CLIA Certificate of Waiver, Certificate of Compliance, or Certificate of Accreditation.- Easy to use POC SARS-CoV-2 Nucleoprotein antigen test that can performed in many settings with symptomatic or asymptomatic patients
- Identifies acute infection in symptomatic patients with high accuracy: PPA (Sensitivity) 86.67%, NPA (Specificity) 100%
- Direct nasal sample
- Results in approximately 20 minutes
- No instrumentation needed
Specifications
15°C to 30°C | |
Waived | |
Direct Nasal Swab | |
SARS-CoV-2 Rapid Antigen Test | |
Intended for the qualitative detection of SARS-CoV-2 Nucleoprotein antigen in direct anterior nasal swab specimens from individuals who are suspected of COVID-19 by their healthcare provider within 5 days of symptom onset or from individuals without symptoms or other epidemiological reasons to suspect COVID-19 when tested twice over two or three days with at least 24 hours and no more than 48 hours between tests. | |
Human | |
20 Min. |
Yes | |
SARS-CoV-2 Antigens | |
0.8667 | |
Yes | |
Pouched cassettes, 1 positive control, 1 negative control, sterile swabs, lysis buffer | |
1 | |
Liquid |
Safety and Handling
ShelfLife : 6 Months
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title