Learn More
Scott Safety™ E-Z Airline Respirator Accessory, Hansen Regulator
For use with Type C pressure demand respirators in non-IDLH environments.
Supplier: Scott Safety™ 7100293249
Description
- E-Z Airline Supplied Air Respirator accessory
- Hansen fitting
- For use with not immediately dangerous to life or health (non-IDLH) applications
Specifications
E-Z Air Line Respirator | |
Hansen supply coupling | |
80444101 | |
Regulator |