Learn More
Hach Company SC200 Universal Controller: 100-240 V AC with one analog pH/ORP/DO sensor input and two 4-20 mA outputs
Supplier: Hach Company LXV404.99.00102
The Hach SC200 Universal Controller is the most versatile controller on the market. It allows the use of both digital and analog sensors, to provide compatibility with the broadest range of sensors. The SC200 replaces the Hach SC100 digital and GLI53 analog controllers with advanced features for easier operator use. Customers may add communication modules for a variety of protocols including MODBUS RTU, Profibus DPV1, and HART. The SC200 is compatible with sensors for over 17 parameters, including pH/ORP, dissolved oxygen, chlorine, conductivity, flow, turbidity, and much more.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.