Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SAS™ Rota Test Controls

For use with SAS Rota Test R-42069020

Supplier:  Thermo Scientific™ R42069101

Catalog No. R42069101

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Explore available promotions

Description

Description

  • Contains 1.0-mL vials: one vial positive and one vial negative control

TRUSTED_SUSTAINABILITY
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.