Learn More
Thermo Scientific Sample and Skimmer Cones for iCAP™ Q/Qnova Series ICP-MS Systems
Supplier: Thermo Scientific 3600812
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Ni Sample cone. 1.1mm diameter orifice for minimal deposition when; analyzing high matrix samples. Solid Ni construction.
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)