Learn More
Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 120mL (4 oz.) 53mm Specimen Containers
Prevent bio-hazardous spills with leak-resistant wide-mouth containers, ideally suited for urine collection from women during pregnancy and for older or overweight patients.
Supplier: Thermo Scientific™ 01 0183
Description
Prevent bio-hazardous spills with the proprietary, all plastic, leak-resistant seal of Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 120 mL (4 oz.) 53mm Specimen Containers. Wide-mouth containers are ideally suited for urine collection from women during pregnancy and for older or overweight patients. These stackable, shatter-resistant containers are manufactured from virgin polypropylene and feature the Bio-Tite™ system. They are tested at varying pressures, making them ideal for use in various transport systems.
- 120mL (4 oz.) 53mm
- Bio-Tite caps create leak-resistant seal for superior safety and handling
- Dual-threaded caps create improved sealing
- Sterilized by gamma irradiation, which eliminates potential contamination
- Top and bottom positive grip knurls are easy to grip and open using latex gloves
- Stackable containers are easy to store, saving lab space
- Virgin polypropylene containers allow higher clarity to see samples, and will not become brittle
- Sterile indicator tab assures tamper evidence
- Bilingual English and Spanish label facilitates accurate usage
Ordering Information:
Case of 300
Specifications
Specimen Container | |
Virgin Polypropylene | |
Non-sterile | |
Blue | |
Bio-Tite Lid | |
53 mm | |
300/Cs. |
Yes | |
Cylindrical | |
120 mL | |
53 mm | |
Cap and container bagged separately | |
No Label | |
300/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.