Learn More
Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 120mL (4 oz.) 53mm Specimen Containers
![GREENER_CHOICE](/content/dam/fishersci/glyphs/greener_choice.png)
Prevent bio-hazardous spills with leak-resistant wide-mouth containers, ideally suited for urine collection from women during pregnancy and for older or overweight patients.
Supplier: Thermo Scientific™ 01 0181
Description
Prevent bio-hazardous spills with the proprietary, all plastic, leak-resistant seal of Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 120 mL (4 oz.) 53mm Specimen Containers. Wide-mouth containers are ideally suited for urine collection from women during pregnancy and for older or overweight patients. These stackable, shatter-resistant containers are manufactured from virgin polypropylene and feature the Bio-Tite™ system. They are tested at varying pressures, making them ideal for use in various transport systems.
- 120 mL (4 oz.) 53 mm
- Bio-Tite caps create leak-resistant seal for superior safety and handling
- Dual-threaded caps create improved sealing
- Sterilized by gamma irradiation, which eliminates potential contamination
- Top and bottom positive grip knurls are easy to grip and open using latex gloves
- Stackable containers are easy to store, saving lab space
- Virgin polypropylene containers allow higher clarity to see samples, and will not become brittle
- Sterile indicator tab assures tamper evidence
- Bilingual English and Spanish label facilitates accurate usage
Ordering Information:
Case of 300
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specimen Container | |
Virgin Polypropylene | |
Non-sterile | |
White | |
Bio-Tite Lid | |
53 mm | |
300/Pk. |
Yes | |
Cylindrical | |
120 mL | |
53 mm | |
Cap and container bagged separately | |
No Label |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.