Learn More
BioMed Diagnostics An isotonic solution typically used for detection of the severe acute respiratory syndrome SARS-CoV-2. | Available in: 50 pack
Supplier: BioMed Diagnostics 11612002
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This Saline Solution (0.85%) has been validated for the detection of the severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) that causes COVID-19 disease in humans. | Product Principle: 1.) Intended to aid in the process of collection and transport of human clinical specimen. 2.) Ensures a non-replicating competent status of SARS-CoV-2. 3.) Is aseptic and does not contain bacteria or fungi growth-inhibition agents. 4.) Validated to be compatible with the identification of SARS-CoV-2 using qRT-PCR technology | Detects: Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2). | Available in: 50 pack
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.