Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Medicom™ SafeMask™ Cone Face Masks
Utility mask made of latex-free, porous material which helps protect the wearer from body fluids and contamination caused by exhaled micro-organisms.
Supplier: Medicom™ 2021
This item is not returnable.
View return policy
Description
- Molded contour design increases comfort and allows for breathability
- Adjustable, aluminum nosepiece and stretchable headband for a custom, close fit
- Materials and construction are soft and non-irritating
- Comfortable to wear and alternative to shingle pleat masks
Specifications
General Purpose Face Mask | |
Headband | |
Standard | |
Cone | |
Dental, industrial, medical, laboratory and safety | |
2021 | |
Shrink wrapped |
Pink | |
Polyester | |
Non-sterile | |
Single-use | |
Yes | |
600 Masks/Cs. | |
SafeMask™ |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction