Learn More
Pyramex™ S1040 Face Shield
Click to view available options
Material: Polycarbonate and Aluminum
Description
Specifications
Certifications/Compliance | ANSI Z87.1 |
Lens Tint | Clear |
For Use With (Equipment) | HGBR |
Weight (English) | 2.7 oz. |
Material | Polycarbonate, Aluminum |