Learn More
Hettich Centrifuges ROTANTA460RF(196)13(48)16,Pk
Supplier: Hettich Centrifuges 460RFBLOODHC
Hettich ROTANTA 460 RF BLOOD TUBE PACKAGE HIGH CAPACITY, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (4) Adapters 13 x 75/100 mm, (4) Adapters 16 x 75/100, (4) Biocontainment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years