Learn More
Hettich Centrifuges ROTANTA460RF,(96)15(40)50,Package
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Hettich Centrifuges 460RFCELLCULTUREHC
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
Hettich ROTANTA 460 RF CELL CULTURE PACKAGE HIGH CAPACITY, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (4) Adapters 15 mL, (4) Adapters 50 mL, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years