Learn More
Mystaire™ Room Air Purifier
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Target fumes at the source or eliminate room air quality issues.
Supplier: Mystaire™ FE1210
Description
- Sparkless and brushless centrifugal fan for quiet operation
- Epoxy coated aluminum construction
- HEPA and/or Carbon Filtration
- HEPA or carbon filter depending on application
- Lightweight and portable
- Fume Extractor: One or two adjustable capture arms for easy movement to the source of contamination
Specifications
Mystaire Ductless Workstation | |
Single-Replacement Capture Arm |