Learn More
IBL America RF-Absorbent (1 mL)
Supplier: IBL America IB79000
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
RF-Absorbent is designed for pre-absorption of interfering substances in IgM assays. Interfering substances may include rheumatoid factors or other serum components capable of causing non-specific signal in ELISA assessment of IgM antibodies. To be used with the appropriate IB79xxx IgM ELISA kits. Please refer to specific ELISA protocols for more information.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.