Learn More
Electron Microscopy Sciences Black Epoxy Thermosetting Hot Mount Resin 1 KG
Supplier: Electron Microscopy Sciences 1213
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Black epoxy with mineral filler, thermosetting. For hard materials. Excellent edge retention.