Learn More
Electron Microscopy Sciences Red bakelite Thermosetting Hot Mount Resin 1 KG
Supplier: Electron Microscopy Sciences 1218
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Red bakelite with wood filler, thermosetting. Routine examination of soft to medium hard materials. Suitable as backing.