Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Replacement Gas Filters
For Restek super clean gas filter kits
Supplier: Restek™ 21982
Description
- High-purity output ensures 99.9999% pure gas (at max. flow of 2 L/min)
- Designed for easy, hassle-free cartridge changes
- Glass inside to prevent diffusion
- Polycarbonate housing outside for safety
- Indicator code shown on every trap to avoid confusion in determining when trap needs to be replaced
Specifications
Max: 2 L/min | |
Restek Super Clean Gas Filter Kits | |
Replacement Gas Fiter |
Helium specific gas filter | |
Removing Oxygen, Moisture and Hydrocarbons |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction