Learn More
Thermo Scientific™ Remel™ Streptex™ Extraction Enzyme
Remel Streptex and Extraction Enzyme are used with Streptex kit.
Supplier: Thermo Scientific™ R30951001
Description
Streptex™ supplementary reagents are intended for use in the streptococcal grouping system provided by the Streptex kit, which contains all components necessary for the system. These reagents are provided as replacement items. For complete information and correct use of this product, the user is referred to the Instructions for Use which accompanies the Streptex kits (ZL50/R30950501 and ZL61/R30164701).
Ordering Alerts:
Freeze dried.
Specifications
Streptex and Extraction Enzyme | |
Streptococcus Test Kit | |
11 mL/Vial, 25 Tests/Kit |
Yes | |
25 Tests/Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.