Learn More
Thermo Scientific™ Remel™ Color Slide™ RF Rapid Latex Test
Latex agglutination slide test for the qualitative and semi-quantitative determination of circulating rheumatoid factor (RF) in human serum
Includes: Positive and negative controls, latex reagent, sample diluent, pipets, and disposable test cards
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
23-033-138A
|
Thermo Scientific™
R2471328 |
40 tests | 40 tests | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
23-033-138B
|
Thermo Scientific™
R2471336 |
100 tests | 100 tests | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- CLIA Complexity: Non-Waived
- CPT Codes: 86431
- Utilizes mono-dispersed latex reagent
- Semi-quantitative titers can be performed directly on the reaction card for efficiency and accuracy
- No sample dilution is required for qualitative procedure
- Positive control is assayed to assist in regulatory compliance to prove proficiency of titering techniques
- Results in uniformly smooth negatives
- Test results within one minute
- For in vitro diagnostic use