Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Remel™ Color Slide™ II Mononucleosis Test
Rapid slide agglutination test using horse erythrocytes for detection of heterophile antibodies of infectious mononucleosis.
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Description | Color Slide™ II Mononucleosis Test |
Detectable Analytes | Mononucleosis Antibodies |
Sample Type | Plasma, Serum |
Type | Reagent |
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity & Availability | ||||
23-046889
|
Thermo Scientific™
R2471088 |
Color Slide™ II Mononucleosis Test | 150 Tests |
Pack of 100 for $837.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-286555
|
Thermo Scientific™
R2468944 |
Color Slide™ II Mononucleosis Test | 50 Tests |
Pack of 50 for $321.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-286554
|
Thermo Scientific™
R2468936 |
Color Slide™ II Mononucleosis Test | 24 Tests |
Pack of 24 for $221.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Thermo Scientific™ Remel Color Slide and II Mononucleosis Test is a rapid slide agglutination test using horse erythrocytes for detection of heterophile antibodies of infectious mononucleosis.
- Dual color indicator
Specifications
2°C to 8°C | |
Mononucleosis Antibodies | |
Reagent |
Color Slide™ II Mononucleosis Test | |
Plasma, Serum |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title