Learn More
Felix Storch Inc REFRIGERATOR-FREEZERS 56.63IN
Supplier: Felix Storch Inc CP962
GENERAL PURPOSE ONLY FOR NON-CRITICAL USE ONLY. Less than 22" wide, the CP962 offers a generous 7.1 cu.ft. storage capacity in a slim fit. This unit comes in a white finish, with a sealed back design to ensure easier cleanup. The door swing is user-reversible, making it easy to adjust the unit to best accommodate your setup.Inside, the CP962 utilizes low maintenance automatic defrost in the refrigerated section and static manual defrost in the freezer.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.