Learn More
Pluriselect Re-Strainer (In-Line-Strainer) 500 µm - Sterile, 4/pack
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Pluriselect 437550040
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Re-Strainer is a filtration device for high volumes of liquid to exclude or concentrate particles from liquids with the possibility of particle recovery. The 6 ml recovery reservoir can be accessed by unscrewing the cap. The Re-Strainer has two female luer-lock-ports.These ports can be used to connect several Re-Strainer to a filter cascade (male to male luer-lock connector) or integrate the Re-Strainer as an in-line-strainer via luer-lock tube adaptor.The mesh is an integrated part of the housing to give the device maximum mechanical stability.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.