Learn More
Ortho-Clinical Diagnostics™ Rare Antisera
![SDP](/content/dam/fishersci/glyphs/sdp.png)
$608.46 - $1699.92
Specifications
For Use With (Application) | For recognition of Antigens on Human red Blood cells |
---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
23-287-461
![]() |
Ortho-Clinical Diagnostics™
721380 |
3 mL | Antisera Anti-Jka BioClone™ |
Each for $998.01
|
|
||||
23-287-462
![]() |
Ortho-Clinical Diagnostics™
723030 |
3 mL | Antisera Anti-S |
Each for $1,649.54
|
|
||||
OK715130
![]() |
Ortho-Clinical Diagnostics™
715130 |
3 mL | Antisera Anti-Fya (Duffy) |
Each for $913.49
|
|
||||
23-287-319
![]() |
Ortho-Clinical Diagnostics™
715850 |
5 mL | Antisera Anti-M (Rabbit) |
Each for $1,336.65
|
|
||||
23-287-322
![]() |
Ortho-Clinical Diagnostics™
715150 |
5 mL | Antisera Anti-Kell |
Each for $608.46
|
|
||||
23-287-323
![]() |
Ortho-Clinical Diagnostics™
721480 |
3 mL | Antisera Anti-Jkb BioClone™ |
Each for $1,699.92
|
|
||||
Description
- Antisera are used as qualitative tests for recognition of antigens on human red blood cells
- Each antisera is used to indicate the presence of specific antigens, which when present will cause clumping of normal human red blood cells
Specifications
For recognition of Antigens on Human red Blood cells |