Learn More
BIOSEARCH TECHNOLOGIES INC RapiDxFire Thermostable Reverse Transcriptase, 250 ReactionsA truly thermostable reverse transcriptase for fast synthesis of short cDNA (< 1 Kb).
Supplier: BIOSEARCH TECHNOLOGIES INC 302502
RapiDxFire Thermostable Reverse Transcriptase, 250 ReactionsA truly thermostable reverse transcriptase for fast synthesis of short cDNA (< 1 Kb).Key features:Extremely active at high temperatures (55 °C to 80 °C): Improves specificity of cDNA Synthesis from diverse RNA templatesSensitive: Detects 100 copies of RNA in two-step RT-qPCR assaysShort reaction time (5 minutes or less): Streamlined RT-qPCR workflow and faster time to resultsStable at room temperature (> 3 months): Simplifies setup on automation decks (no cold storage required) and enables use in environments with limited cold-storageLyo-compatible: Enzyme formulation is free of glycerol -and other components- that are known to interfere with downstream lyophilisationBatch to batch reproducibility: Manufactured in an ISO 13485-certified facility
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.