Learn More
Research Products International Corp Rad-Can II Beta Shield, 3/8 Inch Thick
Supplier: Research Products International Corp BRC15
Features a hinged lid with funnel port. Designed to hold a one gallon polyethylene bottle for storing liquid radioactive waste. Cardboard waste boxes available separately.