Learn More
R&D Systems™ TACS Annexin V-FITC Apoptosis Detection Kit
Facilitates rapid fluorometric quantitation of apoptotic cells
$414.50 - $532.00
Specifications
Product Type | TACS Annexin V-FITC Apoptosis Detection Kit |
---|---|
Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
For Use With (Application) | Facilitates rapid fluorometric quantitation of apoptotic cells |
Includes: TACS Annexin V-FITC, 250uL; 10X Binding Buffer, 20mL; Propidium Iodide, 2.5mL
Catalog Number | Mfr. No. | Kit Contents | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Kit Contents | Quantity | Price | Quantity & Availability | ||||
483001K
|
R&D Systems™
483001K |
TACS Annexin V-FITC, 100μL; 10X Binding Buffer, 8mL; Propidium Iodide, 1mL | 100 Tests |
Each for $414.50
|
|
||||
4830250K
|
R&D Systems™
4830250K |
TACS Annexin V-FITC, 250μL; 10X Binding Buffer, 20mL; Propidium Iodide, 2.5mL | 250 Tests |
Each for $532.00
|
|
||||
Description
Early in the apoptosis process, phosphatidylserine (PS) becomes exposed on the cell surface by flipping from the inner to outer leaflet of the cytoplasmic membrane. This event is thought to be important for macrophage recognition of cells undergoing apopt
Supplied with an optimized binding buffer and propidium iodide, included in the incubation mix to identify cells that have lost membrane integrity
As cells disintegrate, greater access to the inner cell membrane allows for additional Annexin V binding: T
Specifications
TACS Annexin V-FITC Apoptosis Detection Kit | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
Facilitates rapid fluorometric quantitation of apoptotic cells |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.