Learn More
Electron Microscopy Sciences QuantomiX WetSEM QX -302 Calibration Capsule
Supplier: Electron Microscopy Sciences RT58
The QX-302 Calibration Capsule is a QX-302 Capsule designed to assist first time users in finding optimal imaging conditions for WETSEM imaging in their SEM .