Learn More
Electron Microscopy Sciences QuantomiX WetSEM QX -302 AssesSment Kit
Supplier: Electron Microscopy Sciences SK3023
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The QX-302 Capsules are used for imaging various thick, non-adherent samples, such as tissue biopsies, plants and material specimens, in their natural wet state. The capsule is suitable for variable sample sizes, with a maximum diameter of 3 mm and thickness of up to 1 mm. No coating or embedding of the sample are required, enabling electron microscopy imaging with easy sample preparation comparable to light microscopy. Includes 3 QX-302 Capsules and all the necessary accessories.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.