Learn More
Electron Microscopy Sciences QuantomiX WetSEM QX -202 Starter Kit
Supplier: Electron Microscopy Sciences SK20224
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The QX-102 Capsules are used for scanning electron microscope imaging of wet materials and biological samples, including liquid foods, cosmetics, oils, and paints, particles in solutions, adherent and non-adherent human and animal cells, and microorganisms. The samples can be visualized either directly or by following appropriate contrast enhancement staining or labeling procedures. The capsule serves as a cell culture dish, specifically designed for the SEM . No coating or embedding of the sample are required, enabling electron microscopy imaging with easy sample preparation comparable to light microscopy. Includes QX-202 Capsules and all the accessories.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.