Learn More
Brady™ Quik-Align™ 5140 Series Polyester 4 in. Label Legend
Quik-Align™ die-cut numbers and letters are self-spacing and self-aligning: the characters interlock to ensure ideal spacing for professional-looking results every time.
$15.75
Specifications
Material Laminated Polyester
Quantity 10 Labels/Pk.
Adhesive Type Permanent Acrylic
For Use With (Application) General Identification
Height (English) 4.75 in.
View More Specs
Products 8
Catalog Number Mfr. No. Displayed Text Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Displayed Text Width (English) Price Quantity & Availability  
17-200-645
View Documents
Brady™
5140PI
I 0.875 in.
Pack of 10 for $15.75
Add to Cart
17-200-428
View Documents
Brady™
5140PY
Y 3.563 in.
Pack of 10 for $15.75
Add to Cart
17-200-642
View Documents
Brady™
5140PW
W 3.75 in.
Pack of 10 for $15.75
Add to Cart
17-200-580
View Documents
Brady™
5140P0
0 3.063 in.
Pack of 10 for $15.75
Add to Cart
17-200-509
View Documents
Brady™
5140P1
1 1.25 in.
Pack of 10 for $15.75
Add to Cart
17-200-512
View Documents
Brady™
5140P2
2 2.938 in.
Pack of 10 for $15.75
Add to Cart
17-200-583
View Documents
Brady™
5140P3
3 3 in.
Pack of 10 for $15.75
Add to Cart
17-200-586
View Documents
Brady™
5140P4
4 3.25 in.
Pack of 10 for $15.75
Add to Cart
Description
Description
  • Traditional sans serif Gothic font provides excellent legibility even at a distance
  • Card-mounted polyester film characters feature pressure-sensitive adhesive
  • Flexible material conforms to most surfaces and holds up to harsh environments, abrasion, and repeated spills and cleanings
  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (-40°F to +230°F)
Specifications
Specifications
Laminated Polyester
Permanent Acrylic
4.75 in.
-40°F to +230°F
10 Labels/Pk.
General Identification
-40°C to +110°C
Black