Learn More
Thermo Scientific Cyclonic Spray Chamber, Quartz
Supplier:  Thermo Scientific 1317080
Encompass
Quartz cyclonic spray chamber, for use with iCAP Q ICP-MS System, low-volume, baffled cyclonic spray chambers efficiently filter out larger aerosol droplets for improved plasma stability.
Catalog No. NC0515744
Explore available promotions