Learn More
CTL Scientific Supply Corp QUANTOFIX NITRATE/NITRITE
Supplier: CTL Scientific Supply Corp 91313
QUANTOFIX NITRATE/NITRITE 100 strips/pack Test strips for the semi-quantitative determination of nitrate and nitrite in solutions. Gradation of scale: 0-10-25-50-100-250-500 ppm nitrate