Learn More
CTL Scientific Supply Corp QUANTOFIX COPPER
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CTL Scientific Supply Corp 91304
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Test sticks for the semi-quantitative determination of copper (Cu + /Cu 2+ ) in solutions. Gradation of scale: 0-10-30-100-300 mg/l Cu +/2+ 100 strips/box