Learn More
CTL Scientific Supply Corp QUANTOFIX CHROMATE
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CTL Scientific Supply Corp 91301
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Test sticks and reagents for the semi-quantitative determination of chromate in solutions. Gradation of scale: 0-3-10-30-100 mg/l 100 strips/box + reagents