Learn More
CTL Scientific Supply Corp QUANTOFIX ARSENIC
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CTL Scientific Supply Corp 91332
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
QUANTOFIX ARSENIC 100 strips/pack plus reagents Test strips for the semi-quantitative determination of arsenic in solutions. Gradation of scale: 0-0.05-0.1-0.5-1.0-1.7-3.0 ppm As3+/5+