Learn More
Bioassay Systems QuantiChrom™ Iron Assay Kit. For quantitative determination of iron ions Fe3+ and/or Fe2+ and evaluation of drug effects on iron metabolism.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bioassay Systems DIFE250
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For quantitative determination of iron ions Fe3+ and/or Fe2+ and evaluation of drug effects on iron metabolism. Key Features: Sensitive and accurate. Linear detection range 27 µg/dL (4.8 µM) to 1,000 µg/dL (179 µM) iron in 96-well plate assay. Simple and high-throughput. The procedure involves addition of a single working reagent and incubation for 40 min. Can be readily automated as a high-throughput assay for thousands of samples per day. Improved reagent stability and versatility. The optimized formulation has greatly enhanced reagent and signal stability. Cuvette or 96-well plate assay. Low interference in biological samples. No pretreatments are needed. Assays can be directly performed on serum samples. Method: OD590nm. Samples: Biological (e.g. serum) and environmental samples. Species: All. Procedure: Assay takes 30 min. Kit size: 250 tests. Detection limit: 27 µg/dL (4.8 µM).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.