Learn More
Bioassay Systems QuantiChrom™ GTPase Assay Kit. For quantitative determination of ATPase, GTPase or any enzyme activity that liberated free phosphate and high-throughput screen for their inhibitors.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bioassay Systems DATG200
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For quantitative determination of ATPase or GTPase activity and high-throughput screen for their inhibitors. Key Features: High sensitivity. Detection of 0.007 U/L ATPase or GTPase activity. Fast and convenient: single reagent, homogeneous "mix-and-measure" assay allows quantitation of enzyme activity within 30 minutes. Robust and amenable to HTS: detection at 620nm greatly reduces potential interference by colored compounds. Z factors of >0.7 are observed in 96-well and 384-well plates. Can be readily automated on HTS liquid handling systems. Method: OD620nm. Samples: ATPase and GTPase. Species: All. Procedure: Assay takes 30 min. Kit size: 200 tests. Detection limit: 0.007 U/L.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.